Order Entry
Slovakia
ContactUsLinkComponent
Anti-DGAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-DGAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-2332R-FITC
Supplier:  BIOSS INC
Anti-DGAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-2332R-FITC
Supplier:  BIOSS INC

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Diacylglycerol O-acyltransferase 1
  • Antigen symbol:
    DGAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8694
  • Antigen synonyms:
    ARGP1|Acyl coenzyme A|DIAR7|Diglyceride acyltransferase|ARAT|DGAT|EC 2.3.1.20|Diacylglycerol O-acyltransferase 1|ACT-related gene product 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    285-330/488
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol and fatty acyl CoA as substrates. In contrast to DGAT2 it is not essential for survival. May be involved in VLDL (very low density lipoprotein) assembly. In liver, plays a role in esterifying exogenous fatty acids to glycerol. Functions as the major acyl-CoA retinol acyltransferase (ARAT) in the skin, where it acts to maintain retinoid homeostasis and prevent retinoid toxicity leading to skin and hair disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DGAT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat