- Antibody type:Primary
- Antigen name:ZFP36 RING Finger protein-like 1
- Antigen symbol:ZFP36L1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:677
- Antigen synonyms:BRF1|TIS11B|RNF162B|Berg36|cMG1|ERF1|ERF-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
TIS11B is a member of the tristetraprolin family. Tristetraprolin (TTP), or TIS11, is a zinc-binding protein encoded by the immediate-early response gene, Zfp-36. TIS11B, a relative of TTP, localizes to the nucleus and may function as a transcription factor involved in regulating the growth factor response. It is an evolutionarily conserved protein containing two C3H1-type zinc fingers and a repeating cys-his motif. TIS11B is an mRNA binding protein and is known to interact with the 3’-untranslated region of VEGF mRNA, thereby decreasing its stability. This suggests that TIS11B is a potential target in antiangiogenic therapy. In addition, TIS11B may also be an important regulator of myogenesis, as its expression is upregulated during murine myoblast differentiation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZFP36L1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat