Anti-p-AXL Rabbit Polyclonal Antibody [clone: Y703]
Catalog # BWRLBS64021
Supplier: BIOWORLD TECHNOLOGY
Specifications
- Antibody type:Primary
- Antigen symbol:p-AXL
- Clonality:Polyclonal
- Clone:Y703
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human AXL around the phosphorylation site of Tyrosine 703.
p-AXL (Y703) pAb detects endogenous levels of AXL protein only when phosphorylated at Tyr703 (Human) or Tyr697 (Mouse).
Type: Primary
Antigen: p-AXL
Clonality: Polyclonal
Clone: Y703
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat