Anti-p-MEK-4/ MAP2K4 Rabbit Polyclonal Antibody [clone: S80]
Catalog # BWRLBS4167
Supplier: BIOWORLD TECHNOLOGY
Specifications
- Antibody type:Primary
- Antigen symbol:p-MEK-4/ MAP2K4
- Clonality:Polyclonal
- Clone:S80
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human MEK-4 around the phosphorylation site of Serine 80.
p-SEK1/MKK4 (S80) pAb detects endogenous levels of SEK1/MKK4 protein only when phosphorylated at Ser80
Type: Primary
Antigen: p-MEK-4/ MAP2K4
Clonality: Polyclonal
Clone: S80
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat