Anti-mAChR M1 Rabbit Polyclonal Antibody [clone: R336]
Catalog # BWRLBS2708
Supplier: BIOWORLD TECHNOLOGY
Specifications
- Antibody type:Primary
- Antigen symbol:mAChR M1
- Clonality:Polyclonal
- Clone:R336
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 311-360 of Human mAChR M1.
mAChR M1 (R336) pAb detects endogenous levels of mAChR M1 protein.
Type: Primary
Antigen: mAChR M1
Clonality: Polyclonal
Clone: R336
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat