Mini scientific cartridge kit, arium®
171-0978
: VIVA
- Description:Mini scientific kit, arium® ultrapure cartridge
- Seq:110
- Pk:1
Arium® mini scientific pack deionisation cartridge faetures top-down-flow technology which is optimised for the removal of both organic and inorganic constituents. The set was designed specifically to match the unit and delivers ultrapure water that exceeds the ASTM Type 1 quality standard.
- High performance capacity with efficient ion exchange resins
- Fast and effective adsorption of impurities through high-grade activated carbon