Cut resistant gloves, HyFlex® 11-724
: Ansell
These knitted seamless liners with INTERCEPT™ technology yarn and PU coated palm combine excellent flexibility and fit with optimal cut protection.
- Best-in-class cut protection and increased comfort and dexterity
- Palm coating design improves worker grip and extends glove life in abrasive conditions
- Uncoated back allows the hand to breath
- Grey coating hides dirt in environments with dirt exposure
INTERCEPT™ is a specialised knitting technology which blends engineered, synthetic and natural fibres into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Ideal for assembling metal parts and components, light metal presses, maintenance, plastics injection and moulding, and primary assembly.
: EN 388:2016 (4342B); Cat. II
- Catalog No:
- ANSE11-724/9
- ANSE11-724/6
- ANSE11-724/7
- ANSE11-724/11
- ANSE11-724/8
- ANSE11-724/10
- Model:
- 11-724
- 11-724
- 11-724
- 11-724
- 11-724
- 11-724
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Finish:
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
Frequently Bought Together