Order Entry
Slovakia
ContactUsLinkComponent
Human IL-17RC ELISA kit
Human IL-17RC ELISA kit
Catalog # BIRBORB138088-96
Supplier:  BIORBYT
CAS Number:  
undefined
Human IL-17RC ELISA kit
Catalog # BIRBORB138088-96
Supplier:  BIORBYT
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17RC
  • Description:
    Human IL-17RC ELISA Kit
  • Detection range:
    250 - 16000 pg/ml
  • Sensitivity:
    <15 pg/ml

Specifications

About this item

Human IL-17RC ELISA kit for the quantitative detection of human Interleukin-17 receptor C (IL-17RC) in cell culture supernates, serum and plasma (heparin, EDTA).

Interleukin-17 receptor C is a protein that in humans is encoded by the IL17RC gene. By genomic sequence analysis, IL17RC gene was mapped to chromosome 3p25.3-p24.1. This gene encodes a single-pass transmembrane protein that shares limited similarity with the interleukin-17 receptor. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only three have been determined to date.

The biologic activity of IL-17 is dependent on a complex composed of IL-17RA and IL-17RC, suggesting a new paradigm for understanding the interactions between the expanded family of IL-17 ligands and their receptors.