Human recombinant RPLP0 (from insect cells (Sf9))
Supplier: BIORBYT
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:60S acidic ribosomal
- Protein/peptide name:RPLP0
- Purity:Greater than 80% as determined by SDS-PAGE.
- Formulation:RPLP0 is supplied in 20 mM HEPES buffer pH-7.5, 0.01mM EDTA 0.02% SDS.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together