Human Apolipoprotein AI (from E. coli)
Supplier: BIORBYT
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:APOA1 although stable at 4 °C for week, should be stored below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze thaw cycles.
- Protein synonyms:APOA1
- Protein/peptide name:Apolipoprotein AI
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MDEPPQSPWD RVKDLATVYV DVLKDSGRDY VSQFEGSALG KQLNLKLLDN WDSVTSTFSK LREQLGPVTQ EFWDNLEKET EGLRQEMSKD LEEVKAKVQP YLDDFQKKWQ EEMELYRQKV EPLRAELQEG ARQKLHELQE KLSPLGEEMR DRARAHVDAL RTHLAPYSDE LRQRLAARLE ALKENGGARL AEYHAKATEH LSTLSEKAKP ALEDLRQGLL PVLESFKVSF LSALEEYTKK LNTQ.
- Formulation:The APOA1 solution containing 20 mM Tris-HCl buffer (pH8.0) and 10% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together