Human recombinant C1QTNF7 (from E. coli)
: BIORBYT
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:CTRP7
- Protein/peptide name:C1QTNF7
- Purity:Greater than 95% as determined by SDS PAGE.
- Solubility:Add 0.1M Acetate buffer pH4 to prepare working stock solution of approximately 0.5mg/mL and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml
- Sequence:MKHHHHHHAS QPRGNQLKGE NYSPRYICSI PGLPGPPGPP GANGSPGPHG RIGLPGRDGR DGRKGEKGEK GTAGLRGKTG PLGLAGEKGD QGETGKKGPI GPEGEKGEVG PIGPPGPKGD RGEQGDPGLP GVCRCGSIVL KSAFSVGITT SYPEERLPII FNKVLFNEGE HYNPATGKFI CAFPGIYYFS YDITLANKHL AIGLVHNGQY RIKTFDANTG NHDVASGSTV IYLQPEDEVW LEIFFTDQNG LFSDPGWADS LFSGFLLYVD TDYLDSISEDDEL.
- Formulation:Human C1QTNF7 was lyophilized from 30mM Acetate Buffer pH-4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together