Order Entry
Slovakia
ContactUsLinkComponent
HIV-1 p17/24 (from E. coli)
HIV-1 p17/24 (from E. coli)
Supplier:  BIORBYT
Certificates
HIV-1 p17/24 (from E. coli)
Supplier:  BIORBYT

Specifications

  • Source:
    E. coli
  • Storage conditions:
    HIV-1 p17, p24 Protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HIV-1 p17/24
  • Purity:
    Greater than 95.0% as determined by HPLC analysis SDS-PAGE.
  • Formulation:
    1.5 urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X 50% Glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together