Human recombinant Calmodulin (from E. coli)
Supplier: BIORBYT
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Biological activity:Assayed for calcium binding and activation of MLCK.
- Protein/peptide name:Calmodulin
- Purity:Greater than 90% as determined by SDS-PAGE.
- Sequence:MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGYISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQ.
- Formulation:Calmodulin at 1mg/ml in 20 mM HEPES-KOH, pH7, 50mM NaCl, 1mM EDTA and 1mM DTT.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together