Mouse recombinant VEGFR3
Catalog # BIRBORB84236-100
Supplier: BIORBYT
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Protein synonyms:FLT-4
- Protein/peptide name:VEGFR3
- Tested applications:WB , SDS-PAGE