Order Entry
Slovakia
ContactUsLinkComponent
 
Recombinant herpes simplex virus-2 (HSV-2) glycoprotein G (from S. cerevisiae)
  BIRBORB82585-1
 :  BIORBYT
 :  
Recombinant herpes simplex virus-2 (HSV-2) glycoprotein G (from S. cerevisiae)
  BIRBORB82585-1
 :  BIORBYT
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    S. cerevisiae
  • Protein/peptide name:
    Herpes Simplex Virus-2 (HSV-2) Glycoprotein G
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    ELISA , WB