Anti-GLG1 Mouse Monoclonal Antibody [clone: GLG1/970]
Catalog # ABNOMAB14273
Supplier: ABNOVA
Specifications
- Antibody type:Primary
- Antigen name:Golgi glycoprotein 1
- Antigen symbol:GLG1
- Clonality:Monoclonal
- Clone:GLG1/970
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:2734
- Amino acid number:Q92896
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Golgi fraction from human liver cells.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human GLG1.
Type: Primary
Antigen: GLG1
Clonality: Monoclonal
Clone: GLG1/970
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human