Order Entry
Slovakia
ContactUsLinkComponent
Anti-ECM1 Mouse Monoclonal Antibody [clone: ECM1/792]
Anti-ECM1 Mouse Monoclonal Antibody [clone: ECM1/792]
Catalog # ABNOMAB14252
Supplier:  ABNOVA
Anti-ECM1 Mouse Monoclonal Antibody [clone: ECM1/792]
Catalog # ABNOMAB14252
Supplier:  ABNOVA

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Recombinant ECM1
  • Antigen symbol:
    ECM1
  • Clonality:
    Monoclonal
  • Clone:
    ECM1/792
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    1893
  • Amino acid number:
    Q16610
  • Storage buffer:
    In 10 mM PBS.
  • Immunogen:
    Recombinant protein corresponding to full length human ECM1.
  • Purification:
    Protein A/G purification
  • Size:
    100 µG
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant human ECM1.

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: ECM1/792
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human