Anti-KAT3A/CBP Rabbit Polyclonal Antibody
NOVUNB100-381
- Antibody type:Primary
- Antigen symbol:KAT3A/CBP
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Size:100 μg
- Pk:0,1 mg
Rabbit Polyclonal KAT3A/CBP Antibody. Tested Applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Proximity Ligation Assay. Tested Reactivity: Human.
There are 2 scientific publications using this antibody.
Type: Primary
Antigen: KAT3A/CBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human