Anti-Adig Goat Polyclonal Antibody
EVBIEB12488
- Antibody type:Primary
- Antigen symbol:Adig
- Clonality:Polyclonal
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Format:Affinity Purified
- Form:Liquid
- Antigen synonyms:adipogenin|Adig|Smaf1|small adipocyte factor 1|SMAF-1
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:9.36
- Sequence:SQESKEDDSDLC
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:Adipogenin (mouse, aa41-52)
- Purification:Affinity purified
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Type: Primary
Antigen: Adig
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse