Order Entry
Slovakia
ContactUsLinkComponent
Barium chloride dihydrate ≥99.0%, powder ACS
Toxic
Irritant
Barium chloride dihydrate ≥99.0%, powder ACS
Supplier:  MP BIOMEDICALS
Certificates
Toxic
Irritant
Barium chloride dihydrate ≥99.0%, powder ACS
Supplier:  MP BIOMEDICALS

Specification Test Results

  • Form:
    White Granular Powder
  • Storage:
    Store at Room Temperature (15 - 30 °C)
  • Assay:
    ≥99.0%
  • Heavy Metals:
    ≤5 ppm
  • Insoluble Matter:
    ≤0,005%
  • Loss on Drying:
    14,0 - 16,0%
  • pH:
    5,2 - 8,2 (5% aq soln)
  • Oxidizing Substances (as NO3):
    ≤0,005%
  • Calcium:
    ≤0,05%
  • Potassium:
    ≤0,0025%
  • Sodium:
    ≤0,005%
  • Strontium:
    ≤0,1%
  • Iron:
    ≤2 ppm

Specification Test Results

Frequently Bought Together