Chlorhexidine gluconate 20% in aqueous solution
Supplier: MP BIOMEDICALS
Certificates
About this item
CAS Number:18472-51-0
:55-56-1
Formula:C₂₂H₃₀Cl₂N₁₀·2C₆H₁₂O₇
MDL Number:MFCD00083599
:897.76 g/mol
UN:3082
ADR:9,III
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together