Butyric acid ≥99.5%
Catalog # ICNA0210116980
Supplier: MP BIOMEDICALS
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
ADR:8,III
:88.11 g/mol
UN:2820
:GHS06
:GHS05
:GHS07
CAS Number:107-92-6
Formula:C₄H₈O₂
MDL Number:MFCD00002814