Order Entry
Slovakia
ContactUsLinkComponent
 
 
 
Benzyl benzoate 99.0-100.5% (by alkalimetry) Ph. Eur., BP, USP, SAFC®
  1.01806.6025
 :  
 
 
Benzyl benzoate 99.0-100.5% (by alkalimetry) Ph. Eur., BP, USP, SAFC®
  1.01806.6025
 :  

 

  • Pack type:
    Stainless steel drum
  • Pk:
    25 L
  • Seq:
    5327

 

 

  • Assay (by alcalimetric following saponification):
    99.0 - 100.5 %
  • Identity (IR-spectrum):
    passes test
  • Acidity:
    passes test
  • Refractive index (n 20/D):
    1.568 - 1.570
  • Density (Ph Eur, d 20/20):
    1.118 - 1.122
  • Density (USP, d 25/25):
    1.116 - 1.120
  • Solidification point:
    ≥ 18 °C
  • Al (Aluminium):
    ≤ 1 ppm
  • Benzaldehyde:
    ≤ 0.05 %
  • Benzene (HS-GC):
    ≤ 2 ppm
  • Methanol (HS-GC):
    ≤ 3000 ppm
  • Toluene (HS-GC):
    ≤ 890 ppm
  • Other residual solvents (ICH Q3C):
    excluded by production process
  • Sulfated ash:
    ≤ 0.05 %
  • Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1 to 3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.:
    ≤ 0,05 %
  • Corresponds to Ph. Eur., BP, USP:
    ≤ 2 ppm
  • Sulphated Ash:
    ≤ 0,05 

 

 

 :
 :212.25 g/mol
 :324 °C (1013 hPa)
 :21 °C
 :1.12 g/cm³ (20 °C)
 :158 °C
 :Ambient
 :204-402-9
 :3082
 :9,III
 :GHS07
 :GHS09
 :120-51-4
 :C₁₄H₁₂O₂
 :MFCD00003075