Anti-EEF1B2 Mouse Monoclonal Antibody [clone: 3F9]
ABNOH00001933-M12
: ABNOVA
- Antibody type:Primary
- Antigen symbol:EEF1B2
- Clonality:Monoclonal
- Clone:3F9
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Full-length
- Gene ID:1933
- Sequence:MGFGDLKSPAGLQVLNDYLADKSYIEGYVPSQADVAVFEAVSSPPPADLCHALRWYNHIKSYEKEKASLPGVKKALGKYGPADVEDTTGSGATDSKDDDDIDLFGSDDEEESEEAKRLREERLAQYESKKAKKPALVAKSSILLDVKPWDDETDMAKLEECVRSIQADGLVWGSSKLVPVGYGIKKLQIQCVVEDDKVGADMLEEQITAFEDYVQSMDVAAFNKI
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against a full-length recombinant EEF1B2.
Type: Primary
Antigen: EEF1B2
Clonality: Monoclonal
Clone: 3F9
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human,Mouse,Rat