Zadaní objednávky
Slovakia
ContactUsLinkComponent
 
Anti-CD58 Mouse Monoclonal Antibody (FITC) [clone: HI58a]
  ABNOMAB13951
 :  ABNOVA
Anti-CD58 Mouse Monoclonal Antibody (FITC) [clone: HI58a]
  ABNOMAB13951
 :  ABNOVA

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD58
  • Clonality:
    Monoclonal
  • Clone:
    HI58a
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    965
  • Size:
    100 reactions
  • Pk:
    100 Assays

 

 

Mouse monoclonal antibody raised against human CD58.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: HI58a
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human