Anti-ITGB14 Mouse Monoclonal Antibody (PE) [clone: VJ1/14]
Catalog # ABNOMAB13864
Supplier: ABNOVA
Specifications
- Antibody type:Primary
- Antigen symbol:ITGB14
- Clonality:Monoclonal
- Clone:VJ1/14
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:3688
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human ITGB1.
Type: Primary
Antigen: ITGB14
Clonality: Monoclonal
Clone: VJ1/14
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human