Order Entry
Slovakia
ContactUsLinkComponent
Signs, warning stands
  BRDY092110
 :  BRADY
undefined
Signs, warning stands
  BRDY092110
 :  BRADY
  • Description:
    General warning, can be print on
  • Dimensions:
    -
  • Material:
    PP
  • Seq:
    01
  • Pk:
    1
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
  • Made of high-density polypropylene
  • Durable, yellow
  • No buckling or corrosion
  • Large note field is visible from over 9 meters
  • With default notices or unlabeled also available
Height: 660 mm