General purpose gloves, DermiFlex Ultra Glove, A352
: Portwest
These nylon/spandex gloves with PU/nitrile foam coating on palm and fingers are ideal for precision handling.
- 3/4 coated for added protection to the back of the hand
- Seamless breathable knitted liner for comfort and dexterity
EN 388 (3131)
: EN 388:2016 +A1:2018 - 4.1.3.1.X.
- Catalog No:
- 112-4591
- 112-4593
- 112-4592
- Model:
- DermiFlex Ultra Glove, A352
- DermiFlex Ultra Glove, A352
- DermiFlex Ultra Glove, A352
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Liner material:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Finish:
- 3/4 coating (PU/nitrile foam)
- 3/4 coating (PU/nitrile foam)
- 3/4 coating (PU/nitrile foam)
- Gauge:
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388
- EN 388
- EN 388
Frequently Bought Together