Benzamidoxime 97%
:613-92-3
:
:C₇H₈N₂O
:MFCD00031485
:136.15 g/mol
:74…77 °C
:Ambient
:2811
:6.1,III
:GHS06
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- Melting point:73 to 79 °C
- HPLC:≥94.0 %
Frequently Bought Together