
3-Amino-4-iodopyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105752-11-2
:
Formula:C₅H₅IN₂
MDL Number:MFCD04971334
:220.01 g/mol
Melting Pt:65…70 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:60 to 70 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together