Pyrrolidine 99.5+%, AcroSeal®, purified by redistillation
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-75-1
:
Formula:C₄H₉N
MDL Number:MFCD00005249
:71.12 g/mol
Boiling Pt:86…88 °C (760 mmHg)
Melting Pt:–63 °C
Density:0.866 g/cm³
Flash Pt:3 °C
UN:1922
ADR:3,II
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4421 to 1.4441 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥99.4 %
- Color scale:≤10 APHA
Specification Test Results
Related Information
Frequently Bought Together