
Imidazo[1,2-a]pyridine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:274-76-0
:
Formula:C₇H₆N₂
MDL Number:MFCD00005553
:118.1359 g/mol
Boiling Pt:97…98 °C (760 mmHg)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to yellow to brown Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together