Order Entry
Slovakia
ContactUsLinkComponent
n-Nonane, anhydrous 99%, AcroSeal®
Flammable
Irritant
Health
Environment
n-Nonane, anhydrous 99%, AcroSeal®
Catalog # 435631000.
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
n-Nonane, anhydrous 99%, AcroSeal®
Catalog # 435631000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4040 to 1.4060 (20°C, 589 nm)
  • Water:
    ≤0.005 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specification Test Results

About this item

 :
Boiling Pt:151 °C (760 mmHg)
Melting Pt:–53 °C
Density:0.718 g/cm³
Flash Pt:31 °C
Storage Temperature:Ambient
UN:1920
ADR:3,III
 :128.26 g/mol
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:111-84-2
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574