1-Hexylmagnesium chloride 2 M in THF, AcroSeal®
:44767-62-6
:
:C₆H₁₃ClMg
:MFCD06798087
:144.93 g/mol
:65…68 °C (760 mmHg)
:0.965 g/cm³
:–25 °C
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:Ambient
- Appearance:Grey-brown to dark brown Solution
- Titration Grignard:1.90 to 2.20 M
- Melting point:125 to 129 °C
- HPLC:≥97.5 %
Frequently Bought Together