

Ethyltrimethoxysilane 97%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5314-55-6
:
Formula:C₅H₁₄O₃Si
MDL Number:MFCD00051541
:150.25 g/mol
Boiling Pt:123…124 °C (1013 hPa)
Density:0.94 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3830 to 1.3850 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Related Information
Frequently Bought Together