Cyclopropylzinc bromide 0.5 M in THF, AcroSeal®
Catalog # 432820500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Light brown to black Solution
- Titration Grignard:0.30 to 0.60 M
- Water:≤0.5 % (K.F.)
- Anticaking agent:≤0.7 % (Tricalcium phosphate)
- Iron (Fe):≤5 ppm
Specification Test Results
About this item
:
:186.357 g/mol
Density:0.969 g/cm³
Flash Pt:–17 °C
Storage Temperature:Refrigerator
UN:3399
ADR:4.3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:126403-68-7
Formula:C₃H₅BrZn
MDL Number:MFCD08741505