Order Entry
Slovakia
ContactUsLinkComponent
2-Naphthylmagnesium bromide 0.5 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
2-Naphthylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431740500.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Health
2-Naphthylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431740500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Clear brown Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    0.40 to 0.60 M
  • Titration excess NaOH:
    ≥98.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO4):
    ≤0.005 %
  • Hexafluorosilicate (SiF6):
    ≤0.1 %
  • Iron (Fe):
    ≤0.0005 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 % (in water)
  • (NH4)HF2:
    ≤1.0 %

Specification Test Results

About this item

 :
 :231.37 g/mol
Density:0.948 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
 :GHS08
CAS Number:21473-01-8
Formula:C₁₀H₇BrMg
MDL Number:MFCD01319910