Order Entry
Slovakia
ContactUsLinkComponent
Dimethyl sulphate 99+%, AcroSeal®
Toxic
Health
Corrosive
Dimethyl sulphate 99+%, AcroSeal®
Certificates
undefined
Toxic
Health
Corrosive
Dimethyl sulphate 99+%, AcroSeal®
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Oily liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3860 to 1.3880 (20°C, 589 nm)
  • Acidity:
    ≤0.2 % (as Sulfuric acid)
  • Titration:
    99.0 to 101.0 % (Hydrolysis)

Specification Test Results

Frequently Bought Together