Order Entry
Slovakia
ContactUsLinkComponent
p-Toluenesulphonyl isocyanate 96%, AcroSeal®
Irritant
Health
p-Toluenesulphonyl isocyanate 96%, AcroSeal®
Catalog # 430761000.
CAS Number:  
undefined
Irritant
Health
p-Toluenesulphonyl isocyanate 96%, AcroSeal®
Catalog # 430761000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5330 to 1.5360 (20°C, 589 nm)
  • GC:
    ≥95 %
  • Color scale:
    ≤50 APHA
  • Impurity:
    ≤1.1 % Tosyl chloride

Specification Test Results

About this item

 :
 :197.21 g/mol
Boiling Pt:144 °C (10 mmHg)
Melting Pt:–2 °C
Density:1.29 g/cm³
Flash Pt:145 °C
UN:2206
ADR:6.1,III
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:4083-64-1
Formula:C₈H₇NO₃S
MDL Number:MFCD00002030