Acetamidoxime 95+%
:22059-22-9
:MFCD09859749
:C₂H₆N₂O
:MFCD00506021
:74.08 g/mol
:134…138 °C
:GHS07
:Refrigerator
- Appearance:White to off-white Crystals
- Infrared spectrum:Conforms
- Melting point:131 to 138 °C
- Titration with HClO4:≥95.0 %
Frequently Bought Together