1-Hexene 99%, AcroSeal®
:592-41-6
:
:C₆H₁₂
:MFCD00009505
:84.16 g/mol
:62…65 °C (760 mmHg)
:–140 °C
:0.678 g/cm³
:–26 °C
:Ambient
:2370
:3,II
:GHS02
:GHS08
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3867 to 1.3887 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together