



3-Methoxyphenylmagnesium bromide 1 M in THF/toluene, AcroSeal®
Catalog # 426751000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear yellow-brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:0.9 to 1.1 M
- Elemental Analysis:≥92 %
Specification Test Results
About this item
:
:211.341 g/mol
Density:1 g/cm³
Flash Pt:–21 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS08
:GHS05
:GHS02
CAS Number:36282-40-3
Formula:C₇H₇BrMgO
MDL Number:MFCD00672002