Ammonia 7N in methanol, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-41-7
:
Formula:NH₃
MDL Number:MFCD00011418
:17.03 g/mol
Boiling Pt:–33 °C (1013 hPa)
Melting Pt:–77 °C
Density:0.77 g/cm³
Flash Pt:14 °C
UN:3286
ADR:3,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:6.0 to 7.5 N
- Refractive index:1.6110 to 1.6130 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together