
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline) 99%
Catalog # ACRO421920050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light brown to purple Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
Specification Test Results
About this item
:
:293.366 g/mol
Melting Pt:149…150 °C
:GHS07
Storage Temperature:Ambient
CAS Number:23001-29-8
Formula:C₁₉H₁₉NO₂
MDL Number:MFCD00051350