Prostaglandin E1 98+%
Catalog # ACRO235020100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥98.0 %
Specification Test Results
About this item
CAS Number:745-65-3
:
Formula:C₂₀H₃₄O₅
MDL Number:MFCD00077860
:354.487 g/mol
Melting Pt:115…116 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Freezer