Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
Supplier:   GLASSCO
Volumetric flasks, class A, blue markings
Supplier:  GLASSCO
Certificates
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
11 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, clear, with PE stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable at 121 °C
Certifications: According to DIN 12664 and ISO 1042.
Product Family Options
Product Information
  • Capacity
    NS
    Tolerance
    Availability
    Price
  • 5 ml
    10/19
    0,025 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,60 €
    4,60 €
    1 * 1 items
       4,60 €
  • 10 ml
    10/19
    0,025 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,14 €
    5,14 €
    1 * 1 items
       5,14 €
  • 20 ml
    10/19
    0,040 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,00 €
    6,00 €
    1 * 1 items
       6,00 €
  • 25 ml
    10/19
    0,040 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,00 €
    6,00 €
    1 * 1 items
       6,00 €
  • 50 ml
    12/21
    0,060 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,82 €
    6,82 €
    1 * 1 items
       6,82 €
  • 100 ml
    14/23
    0,100 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 8,22 €
    8,22 €
    1 * 1 items
       8,22 €
  • 200 ml
    14/23
    0,150 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 11,39 €
    11,39 €
    1 * 1 items
       11,39 €
  • 250 ml
    14/23
    0,150 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 12,04 €
    12,04 €
    1 * 1 items
       12,04 €
  • 500 ml
    19/26
    0,250 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 14,47 €
    14,47 €
    1 * 1 items
       14,47 €
  • 1000 ml
    24/29
    0,400 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 25,57 €
    25,57 €
    1 * 1 items
       25,57 €
  • 2000 ml
    29/32
    0,600 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 40,69 €
    40,69 €
    1 * 1 items
       40,69 €