Volumetric flasks, class A
Supplier: GLASSCO
Certificates
About this item
Borosilicate glass 3.3, clear, with hollow glass stopper.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- With batch number and Conformity certificate
Certifications: According to DIN 12664 and ISO 1042.
Frequently Bought Together