Mixing measuring cylinders, tall form, class B, borosilicate glass
Supplier: KAVALIER
Certificates
About this item
Borosilicate glass 3.3, class B
- Hexagonal glass base
- Brown graduations
- Avaliable with glass or plastic stopper
Certifications: DIN ISO 4788
Frequently Bought Together