Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type

Sedimentation cones, Imhoff type

Supplier:   KAVALIER
Sedimentation cones, Imhoff type
Supplier:  KAVALIER
Certificates
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type

7 Options Available Below

Product Details & Documents

Borosilicate glass, clear.

  • Autoclavable at 121 °C
  • Available in three versions

Ø×H: 118×470 mm

Product Family Options

With closed tip

Product Information

  • Type
    Availability
    Price
  • VTRB632432430044
    Imhoff cones 1000 ml, graduation in czech
    Imhoff cones 1000 ml, graduation in czech
    with closed tip
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • VTRB632432430144
    Imhoff cones 1000 ml, graduation in german
    Imhoff cones 1000 ml, graduation in german
    with closed tip
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • VTRB632432430244
    Imhoff cones 1000 ml, graduation in swiss
    Imhoff cones 1000 ml, graduation in swiss
    with closed tip
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

With stopcock

Product Information

  • Type
    Availability
    Price
  • VTRB632432440044
    Imhoff cones 1000 ml, graduation in czech
    Imhoff cones 1000 ml, graduation in czech
    with stopcock
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • VTRB632432440144
    Imhoff cones 1000 ml, graduation in german
    Imhoff cones 1000 ml, graduation in german
    with stopcock
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • VTRB632432440244
    Imhoff cones 1000 ml, graduation in swiss
    Imhoff cones 1000 ml, graduation in swiss
    with stopcock
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

With hose connection

Product Information

  • Type
    Availability
    Price
  • VTRB632432450044
    Imhoff cones 1000 ml, graduation in czech
    Imhoff cones 1000 ml, graduation in czech
    with bottom end hose connection
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.