Dibenzyl ketoxime ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1788-31-4
:
Formula:C₁₅H₁₅NO
MDL Number:MFCD00015453
:225.29 g/mol
Melting Pt:122…124 °C
Storage Temperature:Ambient
EINECS:217-251-9
Frequently Bought Together